1,2,3,4-Tetrahydroquinoline ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
B22750.22
B22750.36
B22750.C4
B22750.PG
B22750.A1
B22750.22EA
52
EUR
B22750.22
B22750.36
B22750.C4
B22750.PG
B22750.A1
1,2,3,4-Tetrahydroquinoline ≥99%
1,2,3,4-Tetrahydroquinoline
Purity: 99%
CAS No.: 635-46-1
Molecular Formula: C9H11N
Formula Weight: 133.19
Flash Point: 100°(212°F)
Refractive Index: 1.5950
Melting Point: ca 14°
Boiling Point: 248-250°
Beilstein Reference: 116149
MDL No.: MFCD00006693
Formula:
C₉H₁₁N Boiling Pt: 248…250 °C Melting Pt: ∼ 14 °C Density: 1,059 Flash Pt: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006693 CAS Number: 635-46-1 EINECS: 211-237-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...