1,2,3,4-Tetrahydroquinoline 98%

Supplier: Thermo Scientific
Danger

138270250 138271000 138275000 138270025
ACRO138270250EA 33.5 EUR
ACRO138270250 ACRO138271000 ACRO138275000 ACRO138270025
1,2,3,4-Tetrahydroquinoline 98%
1,2,3,4-Tetrahydroquinoline
Formula: C₉H₁₁N
MW: 133,19 g/mol
Boiling Pt: 249 °C (760 mmHg)
Melting Pt: 15…17 °C
Density: 1,06 g/cm³
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006693
CAS Number: 635-46-1
EINECS: 211-237-6

Order Now

Specification Test Results

Appearance Clear yellow to orange Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5930 to 1.5960 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR