Methyl isobutyrate ≥98%
Supplier: Thermo Scientific
|
Danger
|
A14070.AE
A14070.AP
A14070.AEEA
34.7
EUR
A14070.AE
A14070.AP
Methyl isobutyrate ≥98%
Methyl isobutyrate
Purity: 98%
CAS No.: 547-63-7
Molecular Formula: (CH3)2CHCO2CH3
Formula Weight: 102.13
Flash Point: 3°(37°F)
Refractive Index: 1.3840
Odour: Pleasant
Melting Point: -85°
Boiling Point: 90-92°
Beilstein Reference: 1740720
Merck Reference: 14,6088
UN No.: 1237
MDL No.: MFCD00008914
Formula:
C₅H₁₀O₂ MW: 102,13 g/mol Boiling Pt: 90…92 °C Melting Pt: –85 °C Density: 0,891 Flash Pt: 3 °C (37 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008914 CAS Number: 547-63-7 EINECS: 208-929-5 UN: 1237 ADR: 3,II Merck Index: 14,6088 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...