7-Benzyloxyindole ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: 7-(Benzyloxy)-1H-indole

H25776.14 H25776.06 H25776.03
H25776.14EA 178 EUR
H25776.14 H25776.06 H25776.03
7-Benzyloxyindole ≥98%
7-Benzyloxyindole

Purity: 98% CAS No.: 20289-27-4 Molecular Formula: C15H13NO Formula Weight: 223.28 Melting Point: 70-74° MDL No.: MFCD00037974

Formula: C₁₅H₁₃NO
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00037974
CAS Number: 20289-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR