1-Vinylnaphthalene 95% stabilised
Supplier: Thermo Fisher Scientific
|
Warning
|
Stabilisation:
Stabilised with 4-tert-butylpyrocatechol (TBC)
H51043.03
H51043.06
H51043.03EA
87.8
EUR
H51043.03
H51043.06
1-Vinylnaphthalene 95% stabilised
1-Vinylnaphthalene
Stabilized with 4-tert-butylcatechol
Purity: 95%
CAS No.: 826-74-4
Molecular Formula: C12H10
Formula Weight: 154.21
Storage Temperature: -30°C to -10°C
MDL No.: MFCD00075766
Formula:
C₁₂H₁₀ MW: 154.21 g/mol Boiling Pt: 135…138 °C Melting Pt: 21 Density: 1.04 Storage Temperature: Freezer |
MDL Number:
MFCD00075766 CAS Number: 826-74-4 EINECS: 212-560-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...