3-Indolylglyoxalyl chloride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L00908.09
L00908.18
L00908.09EA
0
EUR
L00908.09
L00908.18
3-Indolylglyoxalyl chloride ≥98%
3-Indolylglyoxalyl chloride
Purity: 98+%
CAS No.: 22980-09-2
Molecular Formula: C10H6ClNO2
Formula Weight: 207.62
Storage Temperature: 2°C to 8°C
Odour: Irritating
Beilstein Reference: 147693
UN No.: 2923
MDL No.: MFCD00015460
Formula:
C₁₀H₆ClNO₂ Melting Pt: ∼ 130 °C (dec.) |
MDL Number:
MFCD00015460 CAS Number: 22980-09-2 EINECS: 245-362-2 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...