4-Chloro-3-sulphamoylbenzoic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
B24057.18
B24057.30
B24057.18EA
59.9
EUR
B24057.18
B24057.30
4-Chloro-3-sulphamoylbenzoic acid ≥98%
4-Chloro-3-sulphamoylbenzoic acid
Purity: 98%
CAS No.: 1205-30-7
Molecular Formula: C7H6ClNO4S
Formula Weight: 235.65
Melting Point: 255-260°
MDL No.: MFCD00012375
Formula:
C₇H₆ClNO₄S MW: 235,65 g/mol Melting Pt: 255…260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012375 CAS Number: 1205-30-7 EINECS: 214-882-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...