3,4,5-Trimethoxyphenol 98.5+%

Supplier: Thermo Fisher Scientific
Warning

189140050 189140250 189141000
ACRO189140050EA 95.7 EUR
ACRO189140050 ACRO189140250 ACRO189141000
3,4,5-Trimethoxyphenol 98.5+%
3,4,5-Trimethoxyphenol
Formula: C₉H₁₂O₄
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00008389
CAS Number: 642-71-7
EINECS: 211-387-2

Order Now

Specification Test Results

Appearance White to yellow to beige to brown Powder or crystals or needles
Infrared spectrum Conforms
Melting point 143 to 149 °C
GC ≥98.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR