Nerol ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
cis-3,7-Dimethyl-2,6-octadien-1-ol
A19808.AE
A19808.AP
A19808.AEEA
70.9
EUR
A19808.AE
A19808.AP
Nerol ≥97%
Nerol
Purity: 97%
CAS No.: 106-25-2
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 77°(171°F)
Refractive Index: 1.4750
Boiling Point: 103-105°/9mm
Beilstein Reference: 1722455
Merck Reference: 14,6475
MDL No.: MFCD00063204
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 103…105 °C (9 mmHg) Density: 0.87 Flash Pt: 77 °C (171 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00063204 CAS Number: 106-25-2 EINECS: 203-378-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...