6-Chlorooxindole ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Chloro-2-oxindole

A19674.06 A19674.14 A19674.22
A19674.06EA 45.3 EUR
A19674.06 A19674.14 A19674.22
6-Chlorooxindole ≥98%
6-Chlorooxindole

Purity: 98% CAS No.: 56341-37-8 Molecular Formula: C8H6ClNO Formula Weight: 167.60 Melting Point: 195-199° MDL No.: MFCD00209962

Formula: C₈H₆ClNO
MW: 167,59 g/mol
Melting Pt: 195…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00209962
CAS Number: 56341-37-8
EINECS: 421-330-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR