2-(4-Chlorophenyl)indole ≥98%
Supplier: Thermo Fisher Scientific
Synonyms:
2-(4-Chlorophenyl)-1H-indole
A13638.06
A13638.03
A13638.06EA
123
EUR
A13638.06
A13638.03
2-(4-Chlorophenyl)indole ≥98%
2-(4-Chlorophenyl)indole
Purity: 98%
CAS No.: 1211-35-4
Molecular Formula: C14H10ClN
Formula Weight: 227.70
Melting Point: 205-207°
MDL No.: MFCD00047159
Formula:
C₁₄H₁₀ClN MW: 227.69 g/mol Melting Pt: 205…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047159 CAS Number: 1211-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...