Adenine sulphate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A16964.09
A16964.18
A16964.09EA
46.2
EUR
A16964.09
A16964.18
Adenine sulphate ≥98%
Adenine sulphate
Purity: 98+%
CAS No.: 321-30-2
Molecular Formula: C10H10N10.H2SO4
Formula Weight: 368.34
Odour: Odourless
Melting Point: ca 210° dec.
Beilstein Reference: 3820263
Merck Reference: 14,152
MDL No.: MFCD00213655
Formula:
C₅H₅N₅·½H₂SO₄ Melting Pt: ∼ 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00213655 CAS Number: 321-30-2 EINECS: 206-286-5 Merck Index: 14,152 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...