1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Ethyl-3-methylimidazolium trifluoromethanesulphonate
L19764.04
L19764.09
L19764.04EA
62.6
EUR
L19764.04
L19764.09
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate ≥98%
1-Ethyl-3-methylimidazolium trifluoromethanesulphonate
Purity: 98+%
CAS No.: 145022-44-2
Molecular Formula: C7H11F3N2O3S
Formula Weight: 260.24
Flash Point: None
Refractive Index: 1.4350
MDL No.: MFCD00210008
Formula:
C₇H₁₁F₃N₂O₃S MW: 260,24 g/mol Density: 1,387 Storage Temperature: Ambient |
MDL Number:
MFCD00210008 CAS Number: 145022-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...