2,3-Dihydrofuran ≥98%
Supplier: Thermo Scientific
|
Danger
|
B20575.14
B20575.22
B20575.36
B20575.14EA
36.2
EUR
B20575.14
B20575.22
B20575.36
2,3-Dihydrofuran ≥98%
2,3-Dihydrofuran
Purity: 98+%
CAS No.: 1191-99-7
Molecular Formula: C4H6O
Formula Weight: 70.09
Storage Temperature: 2°C to 8°C
Flash Point: -24°(-11°F)
Refractive Index: 1.4245
Odour: Pungent
Boiling Point: 54-55°
Beilstein Reference: 103168
UN No.: 3271
MDL No.: MFCD00003205
Formula:
C₄H₆O MW: 70,09 g/mol Boiling Pt: 54…55 °C Density: 0,927 Flash Pt: –24 °C (–11 °F) |
MDL Number:
MFCD00003205 CAS Number: 1191-99-7 EINECS: 214-747-7 UN: 3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...