1-Naphthalenethiol ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
B24876.03
B24876.06
B24876.14
B24876.03EA
40.8
EUR
B24876.03
B24876.06
B24876.14
1-Naphthalenethiol ≥99%
1-Naphthalenethiol
Purity: 99%
CAS No.: 529-36-2
Molecular Formula: C10H8S
Formula Weight: 160.24
MDL No.: MFCD00039599
Formula:
C₁₀H₈S Storage Temperature: Ambient |
MDL Number:
MFCD00039599 CAS Number: 529-36-2 EINECS: 208-462-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...