Thiophenol ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Mercaptobenzene, Phenyl mercaptan, Benzenethiol
A15916.0B
A15916.18
A15916.30
A15916.99
A15916.0BEA
182
EUR
A15916.0B
A15916.18
A15916.30
A15916.99
Thiophenol ≥99%
Thiophenol
Purity: 99+%
CAS No.: 108-98-5
Molecular Formula: C6H6S
Formula Weight: 110.18
Flash Point: 55°(131°F)
Physical Form: Liquid
Density: 1.075
Refractive Index: 1.5900
Sensitivity: Air Sensitive
Appearance: Colourless to pale yellow
Odour: Stench
pH: 5
Melting Point: -15°
Boiling Point: 168-169°
Beilstein Reference: 506523
Merck Reference: 14,9355
UN No.: 2337
MDL No.: MFCD00004826
Formula:
C₆H₆S MW: 110.18 g/mol Boiling Pt: 168…169 °C Melting Pt: –15 °C Density: 1.075 Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004826 CAS Number: 108-98-5 EINECS: 203-635-3 UN: 2337 ADR: 6.1 I Merck Index: 12,09492 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...