Thiophenol for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Mercaptobenzene, Phenyl mercaptan, Benzenethiol
M-Clarity™ quality level = MQ200
8.08159.0005
8.08159.0250
8.08159.1000
8.08159.0005EA
55.7
EUR
8.08159.0005
8.08159.0250
8.08159.1000
Thiophenol for synthesis, Sigma-Aldrich®
Thiophenol
Formula:
C₆H₆S MW: 110.18 g/mol Boiling Pt: 169 Melting Pt: –15 °C Density: 1.0728 g/cm³ (20 °C) Flash Pt: 50 Storage Temperature: Ambient |
MDL Number:
MFCD00004826 CAS Number: 108-98-5 EINECS: 203-635-3 UN: 2337 ADR: 6.1 (3) I Merck Index: 12,09492 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...