4-Pentynylbenzene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Phenyl-1-pentyne

L11608.06 L11608.14
L11608.06EA 93.6 EUR
L11608.06 L11608.14
4-Pentynylbenzene ≥98%
4-Pentynylbenzene

Purity: 98+% CAS No.: 1823-14-9 Molecular Formula: C11H12 Formula Weight: 144.22 Flash Point: 85°(185°F) Refractive Index: 1.5165 Boiling Point: 94°/50mm Beilstein Reference: 2074729 MDL No.: MFCD00039813

Formula: (C₆H₅)CH₂CH₂CH₂CCH
MW: 144.22 g/mol
Boiling Pt: 94 °C (50 mmHg)
Density: 0.950
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039813
CAS Number: 1823-14-9
EINECS: 217-352-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR