Benzyl bromide 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
alpha-Bromotoluene
105871000
105875000
105870010
ACRO105871000EA
70.4
EUR
ACRO105871000
ACRO105875000
ACRO105870010
Benzyl bromide 98%
Benzyl bromide
Formula:
(C₆H₅)CH₂Br MW: 171,04 g/mol Boiling Pt: 199 °C (1013 hPa) Melting Pt: –2 °C Density: 1,43 g/cm³ Flash Pt: 86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000172 CAS Number: 100-39-0 EINECS: 202-847-3 UN: 1737 ADR: 6.1,II Merck Index: 13,01130 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5745 to 1.5765 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...