α,2-Dibromotoluene 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Bromobenzyl bromide
181790250
181791000
ACRO181790250EA
121
EUR
ACRO181790250
ACRO181791000
α,2-Dibromotoluene 98%
α,2-Dibromotoluene
Formula:
C₇H₆Br₂ MW: 249,93 g/mol Boiling Pt: 129 °C (19 mmHg) Melting Pt: 29…32 °C Density: 1,881 g/cm³ (20 °C) Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000173 CAS Number: 3433-80-5 EINECS: 222-334-8 UN: 2923 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.6170 to 1.6190 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...