2-Phenylpropylboronic acid pinacol ester ≥97%

Supplier: Thermo Scientific

Synonyms: 2-Phenyl-1-propylboronic acid pinacol ester, 4,4,5,5-Tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane

H26931.MD H26931.03 H26931.06
H26931.MDEA 0 EUR
H26931.MD H26931.03 H26931.06
2-Phenylpropylboronic acid pinacol ester ≥97%
2-Phenylpropylboronic acid pinacol ester

Purity: 97% CAS No.: 280559-30-0 Molecular Formula: C15H23BO2 Formula Weight: 246.16 MDL No.: MFCD09953475

Formula: C₁₅H₂₃BO₂
MW: 246,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953475
CAS Number: 280559-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR