2-Fluoro-4-pyridineboronic acid pinacol ester 95%
Supplier: Thermo Scientific
|
Warning
|
H27179.03
H27179.06
H27179.03EA
111
EUR
H27179.03
H27179.06
2-Fluoro-4-pyridineboronic acid pinacol ester 95%
2-Fluoro-4-pyridineboronic acid pinacol ester
Purity: 95%
CAS No.: 458532-86-0
Molecular Formula: C11H15BFNO2
Formula Weight: 223.06
Melting Point: 48-52°
MDL No.: MFCD06798253
Formula:
C₁₁H₁₅BFNO₂ MW: 223,05 g/mol Melting Pt: 48…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD06798253 CAS Number: 458532-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...