4-Pyridineboronic acid pinacol ester ≥98%
Supplier: Thermo Scientific
|
Warning
|
L17854.03
L17854.MD
L17854.03EA
147
EUR
L17854.03
L17854.MD
4-Pyridineboronic acid pinacol ester ≥98%
4-Pyridineboronic acid pinacol ester
Purity: 98%
CAS No.: 181219-01-2
Molecular Formula: C11H16BNO2
Formula Weight: 205.07
Melting Point: 150-152°
Beilstein Reference: 7919059
Formula:
C₁₁H₁₆BNO₂ Melting Pt: 150…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD01319051 CAS Number: 181219-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...