3-Bromotoluene 99%
Supplier: Thermo Scientific
|
Warning
|
173410100
173411000
ACRO173410100EA
45.7
EUR
ACRO173410100
ACRO173411000
3-Bromotoluene 99%
3-Bromotoluene
Formula:
C₇H₇Br MW: 171,04 g/mol Boiling Pt: 183,7 °C (1013 hPa) Melting Pt: –40 °C Density: 1,41 g/cm³ Flash Pt: 60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000085 CAS Number: 591-17-3 EINECS: 209-702-3 UN: 1993 ADR: 3,III Merck Index: 13,01424 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5510 to 1.5530 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...