3-Bromotoluene ≥98%
Supplier: Thermo Scientific
|
Warning
|
A13990.14
A13990.22
A13990.36
A13990.14EA
49.1
EUR
A13990.14
A13990.22
A13990.36
3-Bromotoluene ≥98%
3-Bromotoluene
Purity: 98%
CAS No.: 591-17-3
Molecular Formula: C7H7Br
Formula Weight: 171.04
Flash Point: 75°(167°F)
Refractive Index: 1.5520
Melting Point: -40°
Boiling Point: 183-184°
Beilstein Reference: 1903633
Merck Reference: 14,1439
MDL No.: MFCD00000085
Formula:
C₇H₇Br MW: 171,04 g/mol Boiling Pt: 183…184 °C Melting Pt: –40 °C Density: 1,410 Flash Pt: 75 °C (167 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000085 CAS Number: 591-17-3 EINECS: 209-702-3 UN: 1993 ADR: 3,III Merck Index: 14,1439 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...