2-Bromo-α,α,α-trifluorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

A10134.14 A10134.22 A10134.36
A10134.14EA 45.3 EUR
A10134.14 A10134.22 A10134.36
2-Bromo-α,α,α-trifluorotoluene ≥98%
2-Bromo-α,α,α-trifluorotoluene

Flash Point: 68°(154°F)
Refractive Index: 1.4820
Odor: Aromatic
Boiling Point: 167-168°
Beilstein Reference: 1945750

Formula: C₇H₄BrF₃
Boiling Pt: 167…168 °C
Density: 1,662
Flash Pt: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000373
CAS Number: 392-83-6
EINECS: 206-879-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR