α-Bromo-2-nitrotoluene ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
α-Bromo-o-nitrotoluene, 1-(Bromomethyl)-2-nitrobenzene, alpha-Bromo-2-nitrotoluene, 2-Nitrobenzyl bromide
A13127.06
A13127.14
A13127.22
A13127.06EA
54.4
EUR
A13127.06
A13127.14
A13127.22
α-Bromo-2-nitrotoluene ≥98%
α-Bromo-2-nitrotoluene
Purity: 98+%
CAS No.: 3958-60-9
Molecular Formula: C7H6BrNO2
Formula Weight: 216.04
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₇H₆BrNO₂ MW: 216,03 g/mol Melting Pt: 45…48 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007184 CAS Number: 3958-60-9 EINECS: 223-558-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...