4-Butylbenzaldehyde ≥97%

Supplier: Thermo Scientific
Warning

L13438.03 L13438.06
L13438.03EA 0 EUR
L13438.03 L13438.06
4-Butylbenzaldehyde ≥97%
4-Butylbenzaldehyde

Purity: 97% CAS No.: 1200-14-2 Molecular Formula: C11H14O Formula Weight: 162.23 Refractive Index: 1.5260 Melting Point: -43° Boiling Point: 96-98°/5mm Beilstein Reference: 2208267 MDL No.: MFCD00040751

Formula: C₁₁H₁₄O
MW: 162,23 g/mol
Boiling Pt: 96…98 °C (5 mmHg)
Melting Pt: –43 °C
Density: 0,970
Storage Temperature: Ambient
MDL Number: MFCD00040751
CAS Number: 1200-14-2
EINECS: 214-851-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR