Propiophenone 99%

Supplier: Thermo Fisher Scientific

Synonyms: Phenyl ethyl ketone, Ethyl phenyl ketone, 1-Phenyl-1-propanone

131540010 131542500 131540025
ACRO131540010EA 110 EUR
ACRO131540010 ACRO131542500 ACRO131540025
Propiophenone 99%
Propiophenone
Formula: C₆H₅C(O)CH₂CH₃
Boiling Pt: 218 °C (760 mmHg)
Melting Pt: 18 °C
Density: 1 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00009309
CAS Number: 93-55-0
EINECS: 202-257-6
Merck Index: 12,08015
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=796cda47787bc1a939c0285780228746da70cd03&vid=b0290578193d65d6b598ec25da82aff9335f1d45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5280 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR