Benzylacetone 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Phenyl-2-butanone, 4-Phenylbutan-2-one
105830050
105832500
105830010
ACRO105830050EA
26.2
EUR
ACRO105830050
ACRO105832500
ACRO105830010
Benzylacetone 99%
Benzylacetone
Formula:
(C₆H₅)CH₂CH₂C(O)CH₃ MW: 148,2 g/mol Boiling Pt: 235…237 °C (760 mmHg) Melting Pt: -13 °C Density: 0,9875 g/cm³ Flash Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008790 CAS Number: 2550-26-7 EINECS: 219-847-4 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5100 to 1.5130 (20°C, 589 nm) |
GC | ≥98.5 % |
Specific gravity | (25/25°C) 0.985 to 0.99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...