Acetophenone, Supelco®
Supplier: Merck
|
Warning
|
Synonyms:
Methyl phenyl ketone, Phenyl methyl ketone
M-Clarity™ quality level = MQ100
42163-1ML-F
42163-5ML-F
42163-1ML-FEA
26.4
EUR
42163-1ML-F
42163-5ML-F
Acetophenone, Supelco®
Acetophenone
Formula:
C₈H₈O MW: 120,15 g/mol Boiling Pt: 201,7 °C (1013 hPa) Melting Pt: 20 °C Density: 1,027 g/cm³ (20 °C) Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008724 CAS Number: 98-86-2 EINECS: 202-708-7 UN: 3334 ADR: 9, Merck Index: 13,00074 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...