2-Methoxyphenylacetone ≥98%
Supplier: Thermo Fisher Scientific
B24495.03
B24495.06
B24495.03EA
36.1
EUR
B24495.03
B24495.06
2-Methoxyphenylacetone ≥98%
2-Methoxyphenylacetone
Purity: 98+%
CAS No.: 5211-62-1
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point:
Formula:
C₁₀H₁₂O₂ Boiling Pt: 128…131 °C (11 mmHg) Density: 1,062 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008770 CAS Number: 5211-62-1 EINECS: 226-008-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...