3-Methylanthranil acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Amino-3-methylbenzoic acid
B24230.03
B24230.06
B24230.03EA
21.8
EUR
B24230.03
B24230.06
3-Methylanthranil acid 98%
3-Methylanthranil acid
Purity: 98%
CAS No.: 4389-45-1
Molecular Formula: C8H9NO2
Formula Weight: 151.16
Melting Point: 174-177°
Beilstein Reference: 2359694
Formula:
C₈H₉NO₂ MW: 151,16 g/mol Boiling Pt: 315 °C (1013 hPa) Melting Pt: 174…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007745 CAS Number: 4389-45-1 EINECS: 224-505-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...