6-Nitro-o-toluic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Methyl-6-nitrobenzoic acid
L06563.06
L06563.14
L06563.06EA
83.5
EUR
L06563.06
L06563.14
6-Nitro-o-toluic acid ≥98%
6-Nitro-o-toluic acid
Purity: 98%
CAS No.: 13506-76-8
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Melting Point: 153-156°
Beilstein Reference: 2050804
Formula:
C₈H₇NO₄ MW: 181,15 g/mol Boiling Pt: 353 °C (1013 hPa) Melting Pt: 153…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007267 CAS Number: 13506-76-8 EINECS: 236-833-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...