3-Nitro-p-toluic acid 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Methyl-3-nitrobenzoic acid
165990250
165991000
165995000
ACRO165990250EA
29.2
EUR
ACRO165990250
ACRO165991000
ACRO165995000
3-Nitro-p-toluic acid 99%
3-Nitro-p-toluic acid
Formula:
C₈H₇NO₄ MW: 181,15 g/mol Boiling Pt: 360 °C (1013 hPa) Melting Pt: 187…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007174 CAS Number: 96-98-0 EINECS: 202-549-3 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 187 to 190 °C |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...