α,α,α,4-Tetrachlorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Danger

B23446.14 B23446.22
B23446.14EA 25.8 EUR
B23446.14 B23446.22
α,α,α,4-Tetrachlorotoluene ≥98%
α,α,α,4-Tetrachlorotoluene

Purity: 98+% CAS No.: 5216-25-1 Molecular Formula: C7H4Cl4 Formula Weight: 229.92 Flash Point: 140°(284°F) Refractive Index: 1.5730 Melting Point: 5-6° Boiling Point: 245° Beilstein Reference: 1866549 UN No.: 1760 MDL No.: MFCD00000790

Formula: C₇H₄Cl₄
Boiling Pt: 245 °C
Melting Pt: 5…6 °C
Density: 1,495
Flash Pt: 140 °C (284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000790
CAS Number: 5216-25-1
EINECS: 226-009-1
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR