1-Chloro-4-ethylbenzene ≥97%
Supplier: Thermo Fisher Scientific
Warning
|
L07931.06
L07931.14
L07931.06EA
103
EUR
L07931.06
L07931.14
1-Chloro-4-ethylbenzene ≥97%
1-Chloro-4-ethylbenzene
Purity: 97%
CAS No.: 622-98-0
Molecular Formula: C8H9Cl
Formula Weight: 140.61
Flash Point: 62°(144°F)
Refractive Index: 1.5175
Odour: Aromatic
Boiling Point: 184-185°
Beilstein Reference: 1099335
MDL No.: MFCD00040855
Formula:
C₈H₉Cl Boiling Pt: 184…185 °C Density: 1,030 Flash Pt: 62 °C (144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040855 CAS Number: 622-98-0 EINECS: 210-763-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...