2-Chloro-4-fluorotoluene ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Chloro-4-fluoro-1-methylbenzene
B20278.14
B20278.22
B20278.36
B20278.14EA
34
EUR
B20278.14
B20278.22
B20278.36
2-Chloro-4-fluorotoluene ≥98%
2-Chloro-4-fluorotoluene
Purity: 98%
CAS No.: 452-73-3
Molecular Formula: C7H6ClF
Formula Weight: 144.58
Flash Point: 50°(122°F)
Refractive Index: 1.4990
Odour: Aromatic
Boiling Point: 154-156°
Beilstein Reference: 1931690
UN No.: 1993
MDL No.: MFCD00000572
Formula:
C₇H₆ClF MW: 144,58 g/mol Boiling Pt: 154…156 °C Density: 1,198 Flash Pt: 50 °C (122 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000572 CAS Number: 452-73-3 EINECS: 207-209-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...