α-Chloro-4-nitrotoluene ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Nitrobenzyl chloride, 4-Nitrobenzylchloride, α-Chloro-p-nitrotoluene

A15749.09 A15749.18 A15749.30
A15749.09EA 0 EUR
A15749.09 A15749.18 A15749.30
α-Chloro-4-nitrotoluene ≥99%
α-Chloro-4-nitrotoluene

Purity: 99% CAS No.: 100-14-1 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: 70-74° Beilstein Reference: 387187 UN No.: 3261 MDL No.: MFCD00007374

Formula: C₇H₆ClNO₂
MW: 171,58 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 70…74 °C
Density: 1,5647 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007374
CAS Number: 100-14-1
EINECS: 202-822-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR