Benzotrifluoride 99+%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
α,α,α-Trifluorotoluene, Solvent BTF
139770500
139775000
139770025
ACRO139770500EA
26.2
EUR
ACRO139770500
ACRO139775000
ACRO139770025
Benzotrifluoride 99+%
Benzotrifluoride
Formula:
C₇H₅F₃ MW: 146,11 g/mol Boiling Pt: 102 °C (760 mmHg) Melting Pt: –29 °C Density: 1,19 g/cm³ Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000372 CAS Number: 98-08-8 EINECS: 202-635-0 UN: 2338 ADR: 3,II Merck Index: 13,01111 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4130 to 1.4150 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...