α,α,α-Trifluoro-2-iodotoluene ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 99%
CAS No.: 444-29-1
Molecular Formula: C7H4F3I
Formula Weight: 272.01
Flash Point: 81°(177°F)
Physical Form: Liquid
Density: 1.936
Refractive Index: 1.5305
Sensitivity: Light Sensitive
Appearance: Colourless to pale yellow
Boiling Point: 198-199°
Beilstein Reference: 2090038
MDL No.: MFCD00001041
Formula:
C₇H₄F₃I Boiling Pt: 198…199 °C Density: 1,936 Flash Pt: 81 °C (177 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001041 CAS Number: 444-29-1 EINECS: 207-147-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...