α,α,α-Trifluoro-3-iodotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: α,α,α-Trifluoro-m-iodotoluene, 3-Iodobenzotrifluoride, 1-Iodo-3-(trifluoromethyl)benzene

A11843.09 A11843.18 A11843.30
A11843.09EA 36.6 EUR
A11843.09 A11843.18 A11843.30
α,α,α-Trifluoro-3-iodotoluene ≥98%
α,α,α-Trifluoro-3-iodotoluene

Purity: 98+% CAS No.: 401-81-0 Molecular Formula: C7H4F3I Formula Weight: 272.01 Flash Point: 70°(158°F) Refractive Index: 1.5185 Melting Point: -8° Boiling Point: 81-82°/24mm Beilstein Reference: 2088596 MDL No.: MFCD00001049

Formula: C₇H₄F₃I
MW: 272.01 g/mol
Boiling Pt: 81…82 °C (24 mmHg)
Melting Pt: –8 °C
Density: 1.882
Flash Pt: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001049
CAS Number: 401-81-0
EINECS: 206-934-7
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR