3-Iodotoluene ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Iodo-3-methylbenzene

A10999.18 A10999.22 A10999.30
A10999.18EA 79.5 EUR
A10999.18 A10999.22 A10999.30
3-Iodotoluene ≥99%
3-Iodotoluene

Purity: 99% CAS No.: 625-95-6 Molecular Formula: C7H7I Formula Weight: 218.04 Flash Point: 82°(179°F) Refractive Index: 1.6040 Melting Point: -27° Boiling Point: 80-82°/10mm Beilstein Reference: 1903634 MDL No.: MFCD00001050

Formula: C₇H₇I
MW: 218.04 g/mol
Boiling Pt: 80…82 °C (10 mmHg)
Melting Pt: –27 °C
Density: 1.698
Flash Pt: 82 °C (179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001050
CAS Number: 625-95-6
EINECS: 210-918-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR