4-Decylaniline ≥98%

Supplier: Thermo Fisher Scientific
Warning

L08620.03 L08620.06 L08620.14
L08620.03EA 43.5 EUR
L08620.03 L08620.06 L08620.14
4-Decylaniline ≥98%
4-Decylaniline

Purity: 98% CAS No.: 37529-30-9 Molecular Formula: C16H27N Formula Weight: 233.40 Flash Point: 110°(230°F) Refractive Index: 1.5100 Melting Point: 23-25° Boiling Point: 218-220°/14mm Beilstein Reference: 2721403 MDL No.: MFCD00007918

Formula: C₁₆H₂₇N
MW: 233.4 g/mol
Boiling Pt: 218…220 °C (14 mmHg)
Melting Pt: 23…25 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007918
CAS Number: 37529-30-9
EINECS: 253-546-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR