3-tert-Butylphenol 99+%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
m-tert-Butylphenol
154380250
154381000
154380010
ACRO154380250EA
130
EUR
ACRO154380250
ACRO154381000
ACRO154380010
3-tert-Butylphenol 99+%
3-tert-Butylphenol
Formula:
C₁₀H₁₄O MW: 150,22 g/mol Boiling Pt: 125…130 °C (20 mmHg) Melting Pt: 40…43 °C Density: 1,048 g/cm³ (20 °C) Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002300 CAS Number: 585-34-2 EINECS: 209-553-4 UN: 2923 ADR: 8,III |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 40 to 43 °C |
GC | ≥99.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...