Butylbenzene 99+%

Supplier: Thermo Scientific
Warning

107850500 107852500 107850010
ACRO107850500EA 64.4 EUR
ACRO107850500 ACRO107852500 ACRO107850010
Butylbenzene 99+%
Butylbenzene
Formula: C₁₀H₁₄
Boiling Pt: 183 °C (760 mmHg)
Melting Pt: –88 °C
Density: 0,86 g/cm³
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00009463
CAS Number: 104-51-8
EINECS: 203-209-7
UN: 2709
ADR: 3,III
Merck Index: 13,01548

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4880 to 1.4900 (20°C, 589 nm)
GC ≥99.0 %
Methanol ≤0.5 % (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR