Dodecylbenzene ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: n-Dodecylbenzene, 1-Phenyldodecane

B21362.06 B21362.14 B21362.22
B21362.06EA 45.5 EUR
B21362.06 B21362.14 B21362.22
Dodecylbenzene ≥97%
Dodecylbenzene

Purity: 97% CAS No.: 123-01-3 Molecular Formula: C18H30 Formula Weight: 246.44 Flash Point: 124°(255°F) Refractive Index: 1.4820 Melting Point: -7° Boiling Point: 331° Beilstein Reference: 1909107 MDL No.: MFCD00008974

Formula: C₁₈H₃₀
MW: 246.44 g/mol
Boiling Pt: 331 °C
Melting Pt: –7 °C
Density: 0.856
Flash Pt: 124 °C (255 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008974
CAS Number: 123-01-3
EINECS: 204-591-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR