Octylbenzene 99%

Supplier: Thermo Fisher Scientific

150990250 150991000
ACRO150990250EA 98.5 EUR
ACRO150990250 ACRO150991000
Octylbenzene 99%
Octylbenzene
Formula: C₁₄H₂₂
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: –36 °C
Density: 0,85 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00009564
CAS Number: 2189-60-8
EINECS: 218-582-1

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4850 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR