Chloro(diphenyl)methane ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Benzhydryl chloride

B24764.14 B24764.22 B24764.36
B24764.14EA 66.4 EUR
B24764.14 B24764.22 B24764.36
Chloro(diphenyl)methane ≥98%
Chloro(diphenyl)methane

Purity: 98% CAS No.: 90-99-3 Molecular Formula: C13H11Cl Formula Weight: 202.69 Flash Point: 160°(320°F) Refractive Index: 1.5950 Odour: Pungent Melting Point: 17-20° Boiling Point: 139-140°/3mm Beilstein Reference: 607925 UN No.: 3265 MDL No.: MFCD00000855

Formula: (C₆H₅)₂CHCl
Boiling Pt: 139…140 °C (3 mmHg)
Melting Pt: 17…20 °C
Density: 1,14
Flash Pt: 160 °C (320 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000855
CAS Number: 90-99-3
EINECS: 202-031-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR