Tribenzylamine ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
B23797.14
B23797.22
B23797.14EA
23.9
EUR
B23797.14
B23797.22
Tribenzylamine ≥99%
Tribenzylamine
Purity: 99+%
CAS No.: 620-40-6
Molecular Formula: (C6H5CH2)3N
Formula Weight: 287.41
Flash Point: 203°(397°F)
Odour: Weak
Boiling Point: 230°/13mm
Beilstein Reference: 2214682
MDL No.: MFCD00004773
Formula:
N(CH₂C₆H₅)₃ MW: 287.4 g/mol Boiling Pt: 230 °C (13 mmHg) Melting Pt: 91…94 °C Density: 0.9912 g/cm³ (95 °C) Flash Pt: 203 °C (397 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004773 CAS Number: 620-40-6 EINECS: 210-638-3 UN: 1325 ADR: 4.1 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...